3-Methoxy-o-phenylenediamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methoxybenzene-1,2-diamine
H35286-03
H35286-MD
AAH35286-03EA
366.56
CAD
AAH35286-03
AAH35286-MD
3-Methoxy-o-phenylenediamine 97%
3-Methoxy-o-phenylenediamine
Purity: 97%
CAS No.: 37466-89-0
Molecular Formula: C7H10N2O
Formula Weight: 138.17
Storage Temperature: -30°C to -10°C
Melting Point: 39-40°
UN No.: 2811
MDL No.: MFCD08276903
Formula:
C₇H₁₀N₂O Melting Pt: 39…40 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08276903 CAS Number: 37466-89-0 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...