3-(Pentafluorothio)aniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Aminophenylsulphurpentafluoride, 3-Aminophenylsulphur pentafluoride, 3-Aminophenylsulfur pentafluoride, 3-Aminophenylsulfurpentafluoride
H35190-03
H35190-06
AAH35190-03EA
113.29
CAD
AAH35190-03
AAH35190-06
3-(Pentafluorothio)aniline 97%
3-(Pentafluorothio)aniline
Purity: 97%
CAS No.: 2993-22-8
Molecular Formula: C6H5F5NS
Formula Weight: 218.17
UN No.: 2811
MDL No.: MFCD00674199
Formula:
C₆H₆F₅NS MW: 218.17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00674199 CAS Number: 2993-22-8 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...