Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H35174-03
H35174-MD
AAH35174-03EA
206.98
CAD
AAH35174-03
AAH35174-MD
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride 95%
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Purity: 95%
CAS No.: 859833-15-1
Molecular Formula: C8H6Cl2N2O
Formula Weight: 217.06
Melting Point: 248-250°
UN No.: 3261
MDL No.: MFCD07772809
Formula:
C₈H₆Cl₂N₂O Melting Pt: 248…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD07772809 CAS Number: 859833-15-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...