2-Aminooxazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34360-03
H34360-06
AAH34360-03EA
230.68
CAD
AAH34360-03
AAH34360-06
2-Aminooxazole 97%
2-Aminooxazole
Purity: 97%
CAS No.: 4570-45-0
Molecular Formula: C3H4N2O
Formula Weight: 84.10
Melting Point: 93-95°
MDL No.: MFCD07364485
Formula:
C₃H₄N₂O Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD07364485 CAS Number: 4570-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...