6-(1H-pyrazol-1-yl)nicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34104-03
H34104-MD
AAH34104-03EA
104.75
CAD
AAH34104-03
AAH34104-MD
6-(1H-pyrazol-1-yl)nicotinic acid 97%
6-(1H-pyrazol-1-yl)nicotinic acid
Purity: 97%
CAS No.: 253315-22-9
Molecular Formula: C9H7N3O2
Formula Weight: 189.20
Melting Point: 272-274°
MDL No.: MFCD00140749
Formula:
C₉H₇N₃O₂ Melting Pt: 272…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD00140749 CAS Number: 253315-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...