Methyl-3-aminoisonicotinate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-aminoisonicotinate, Methyl 3-aminopyridine-4-carboxylate

H34007-03 H34007-09
AAH34007-03EA 123.87 CAD
AAH34007-03 AAH34007-09
Methyl-3-aminoisonicotinate 97%
Methyl 3-aminoisonicotinate

Purity: 97% CAS No.: 55279-30-6 Molecular Formula: C7H8N2O2 Formula Weight: 152.20 Melting Point: 84-86° MDL No.: MFCD02082535

Formula: C₇H₈N₂O₂
MW: 152.20 g/mol
Melting Pt: 84…86 °C
Storage Temperature: Ambient
MDL Number: MFCD02082535
CAS Number: 55279-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR