6-nitro-3,4-dihydro-2H-1,4-benzoxazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H33900-03 H33900-MD
AAH33900-03EA 138.72 CAD
AAH33900-03 AAH33900-MD
6-nitro-3,4-dihydro-2H-1,4-benzoxazine 97%
6-nitro-3,4-dihydro-2H-1,4-benzoxazine

Purity: 97% CAS No.: 28226-22-4 Molecular Formula: C8H8N2O3 Formula Weight: 180.20 Melting Point: 117-119° MDL No.: MFCD07698591

Formula: C₈H₈N₂O₃
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD07698591
CAS Number: 28226-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR