1-Octadecyne 97+%

Supplier: Thermo Scientific Chemicals

H33288-03 H33288-09
AAH33288-03EA 106.49 CAD
AAH33288-03 AAH33288-09
1-Octadecyne 97+%
1-Octadecyne

Purity: 97+% CAS No.: 629-89-0 Molecular Formula: C18H34 Formula Weight: 250.47 Melting Point: 21-24° Boiling Point: 313° MDL No.: MFCD00015088

Formula: C₁₈H₃₄
Boiling Pt: 313 °C
Melting Pt: 21…24 °C
Density: 0.803
Storage Temperature: Ambient
MDL Number: MFCD00015088
CAS Number: 629-89-0
EINECS: 211-114-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR