3-Amino-5-bromo-1-methyl-1H-indazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33202-06
H33202-MD
H33202-03
CAAAH33202-06EA
669.92
CAD
CAAAH33202-06
CAAAH33202-MD
AAH33202-03
3-Amino-5-bromo-1-methyl-1H-indazole 97%
3-Amino-5-bromo-1-methyl-1H-indazole
Purity: 97%
CAS No.: 1000018-06-3
Molecular Formula: C8H8BrN3
Formula Weight: 226.08
Melting Point: 133-135°
MDL No.: MFCD09864804
Formula:
C₈H₈BrN₃ MW: 226.08 g/mol Melting Pt: 133…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD09864804 CAS Number: 1000018-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...