8-Amino-2-naphthol 98%

Supplier: Thermo Scientific Chemicals
Warning

A19404-03 A19404-06 A19404-14
CAAAA19404-03EA 34.76 CAD
CAAAA19404-03 CAAAA19404-06 AAA19404-14
8-Amino-2-naphthol 98%
8-Amino-2-naphthol

Purity: 98% CAS No.: 118-46-7 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 206-210° MDL No.: MFCD00004031

Formula: C₁₀H₉NO
Melting Pt: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004031
CAS Number: 118-46-7
EINECS: 204-252-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR