2-Amino-6-methylpyridine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-Methyl-2-pyridylamine, 6-Methylpyridin-2-amine, 2-Amino-6-picoline
A10444-22
A10444-36
CAAAA10444-22EA
66.83
CAD
CAAAA10444-22
CAAAA10444-36
2-Amino-6-methylpyridine 99%
2-Amino-6-methylpyridine
Purity: 99%
CAS No.: 1824-81-3
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 103°(217°F)
Odor: Unpleasant
Boiling Point: 208-209°
Beilstein Reference: 107048
UN No.: 2811
MDL No.: MFCD00006331
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 208…209 °C Melting Pt: 41…45 °C Flash Pt: 103 °C(217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006331 CAS Number: 1824-81-3 EINECS: 217-360-1 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...