(4-Methoxyphenyl)magnesium bromide 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Anisylmagnesium bromide

089435-AE 089435-AP
AA89435-AEEA 152.5 CAD
AA89435-AE AA89435-AP
(4-Methoxyphenyl)magnesium bromide 0.5 M in THF
(4-Methoxyphenyl)magnesium bromide

Molecular Formula: C7H7BrMgO Formula Weight: 211.34 Flash Point: -20°(-4°F) Physical Form: Liquid Density: 0.955 Sensitivity: Air and Moisture Sensitive Appearance: Brown UN No.: 2924 MDL No.: MFCD00013515

Formula: C₇H₇BrMgO
MW: 211.34 g/mol
Density: 0.955
Flash Pt: –20 °C (–4 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013515
CAS Number: 13139-86-1
UN: 2924
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR