Phenylmagnesium bromide 3 M Stabilised with diethyl ether
Supplier: Thermo Scientific Chemicals
|
Danger
|
087326-AE
087326-AP
AA87326-AEEA
148.85
CAD
AA87326-AE
AA87326-AP
Phenylmagnesium bromide 3 M Stabilised with diethyl ether
Phenylmagnesium bromide
CAS No.: 100-58-3
Molecular Formula: C6H5MgBr
Formula Weight: 181.31
Physical Form: Liquid
Density: 1.14
Sensitivity: Light, Air and Moisture Sensitive
Appearance: Yellow to brown
Odor: Ether-like
UN No.: 3399
MDL No.: MFCD00000038
Grignard reagent
Formula:
C₆H₅BrMg MW: 181.31 g/mol Density: 1.14 Storage Temperature: Ambient |
MDL Number:
MFCD00000038 CAS Number: 100-58-3 UN: 3399 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...