Strontium iodide, anhydrous ≥99%

Supplier: Thermo Scientific Chemicals
Warning

047268-06 047268-14 047268-22
AA47268-06EA 93.85 CAD
AA47268-06 AA47268-14 AA47268-22
Strontium iodide, anhydrous ≥99%
Strontium iodide

Purity: 97% CAS No.: 10476-86-5 Molecular Formula: SnI2 Formula Weight: 341.43 Physical Form: Powder Density: 4.550 Appearance: Pale yellow to dark brown Odor: Odorless Melting Point: 515° Merck Reference: 14,8844 MDL No.: MFCD00049557

Formula: I₂Sr
MW: 341.43
Melting Pt: 515
Density: 4.550
Storage Temperature: Ambient
MDL Number: MFCD00049557
CAS Number: 10476-86-5
EINECS: 233-972-1
UN: 3260
ADR: 8 II
Merck Index: 14,8844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR