D-(+)-Glucose monohydrate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
047263-0E
047263-36
AA47263-0EEA
139.04
CAD
AA47263-0E
AA47263-36
D-(+)-Glucose monohydrate 99%
D(+)-Glucose monohydrate
Purity: 99%
CAS No.: 14431-43-7
Molecular Formula: C6H12O6.H2O
Formula Weight: 198.18 (180.16anhy)
Physical Form: Powder
Appearance: White
Melting Point: ca 83°
Beilstein Reference: 3694181
MDL No.: MFCD00149450
Formula:
C₆H₁₂O₆·H₂O MW: 198.17 g/mol Melting Pt: 83 Storage Temperature: Ambient |
MDL Number:
MFCD00149450 CAS Number: 14431-43-7 EINECS: 200-075-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...