Aluminium oxide, α-phase ≥99.997% (metals basis)
Supplier: Thermo Scientific Chemicals
047261-14
047261-22
047261-36
AA47261-14EA
112.97
CAD
AA47261-14
AA47261-22
AA47261-36
Aluminium oxide, α-phase ≥99.997% (metals basis)
Aluminium oxide, α-phase
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 40 Micron Crystalline
Density: 3.965
Appearance: White
Odor: Odorless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ Boiling Pt: 2980 °C Melting Pt: 2045 °C Density: 3.965 Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...