Calcium carbonate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
047235-06
047235-14
047235-22
AA47235-06EA
93.69
CAD
AA47235-06
AA47235-14
AA47235-22
Calcium carbonate ≥99.999% (metals basis), Puratronic®
Calcium carbonate
Purity: 99.999%
CAS No.: 471-34-1
Molecular Formula: CaCO3
Formula Weight: 100.09
Physical Form: Powder
Density: 2.930
Appearance: White
Odor: Odorless
Melting Point: 800° dec.
Merck Reference: 14,1657
MDL No.: MFCD00010906
Formula:
CaCO₃ Melting Pt: 800 °C (dec.) Density: 2.930 Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 14,1657 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...