Triiron aluminide ≥97.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

047233-18 047233-30
AA47233-18EA 157.89 CAD
AA47233-18 AA47233-30
Triiron aluminide ≥97.5% (metals basis)
Triiron aluminide

Purity: 97.5% CAS No.: 12003-42-8 Molecular Formula: AlFe3 Physical Form: -100 Mesh Powder, Intermetallic Density: 6.72 Appearance: Grey Melting Point: 1540° MDL No.: MFCD00210617

Formula: AlFe₃
Melting Pt: 1540 °C
Density: 6.72
Storage Temperature: Ambient
MDL Number: MFCD00210617
CAS Number: 12003-42-8
EINECS: 234-427-0
UN: 3178
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR