Zirconium hydride ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zirconium(Ⅱ)bydride, Dihydridezirconium(II), Tetrahydridozirconium
047203-09
047203-18
047203-30
AA47203-09EA
58.05
CAD
AA47203-09
AA47203-18
AA47203-30
Zirconium hydride ≥99%
Zirconium hydride
Purity: 99%
CAS No.: 7704-99-6
Molecular Formula: ZnH2
Physical Form: -325 Mesh Powder
Sensitivity: Moisture Sensitive
Odor: Odorless
Merck Reference: 14,10176
UN No.: 1437
MDL No.: MFCD00148878
Formula:
H₄Zr MW: 95.256 Da Storage Temperature: Ambient |
MDL Number:
MFCD00148878 CAS Number: 7704-99-6 EINECS: 231-727-3 UN: 1437 ADR: 4.1,II Merck Index: 14,10176 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...