Diyttrium trioxide ≥99.995%, nanopowder

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Yttrium oxide, Yttrium (III) oxide

047154-14 047154-22 047154-36
AA47154-14EA 127.03 CAD
AA47154-14 AA47154-22 AA47154-36
Diyttrium trioxide ≥99.995%, nanopowder
Diyttrium trioxide

Purity: 99.995% CAS No.: 1314-36-9 Molecular Formula: O3Y2 Formula Weight: 225.81 Physical Form: 50-70nm APS Powder Density: 5.01 Appearance: White Odor: Odorless Melting Point: 2410° Merck Reference: 14,10107 MDL No.: MFCD00011473

Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300
Melting Pt: 2410
Density: 5.01
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR