Viscosity Standard, Specpure®, Nominal Values at 40 and 100 °C, Thermo Scientific
Supplier: Thermo Scientific Chemicals
047013-AP
047036-AP
047100-AP
047158-AP
047021-AP
047033-AP
AA47013-APEA
188.67
CAD
AA47013-AP
AA47036-AP
AA47100-AP
AA47158-AP
AA47021-AP
AA47033-AP
Viscosity Standard, Specpure®, Nominal Values at 40 and 100 °C, Thermo Scientific
Standards
Viscometry Standards
Physical Form: Liquid
MDL No.: MFCD00084411
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...