Platinum ≥99.98% (metals basis), powder -200/+400 mesh

Supplier: Thermo Scientific Chemicals

46830-03 46830-06 046830-03 046830-06
CAAA46830-03EA 494.54 CAD
CAAA46830-03 CAAA46830-06 CAAA46830-MC AA46830-03 AA46830-06 AA46830-MC
Platinum ≥99.98% (metals basis), powder -200/+400 mesh
Platinum

Purity: 99.98% CAS No.: 7440-06-4 Odor: Odorless MDL No.: MFCD0001117

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR