Tripotassium orthophosphate ≥98.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
046644-30
046644-A1
046644-A7
AA46644-30EA
138.53
CAD
AA46644-30
AA46644-A1
AA46644-A7
Tripotassium orthophosphate ≥98.0% ACS
tri-Potassium orthophosphate
Purity: 98.0% min
CAS No.: 7778-53-2
Molecular Formula: K3PO4
Formula Weight: 212.27
Physical Form: Granular
Density: 2.564
Appearance: White
Melting Point: 1340°
Solubility: Soluble in water. Insoluble in alcohol.
Merck Reference: 14,7660
MDL No.: MFCD00036295
Formula:
K₃O₄P Melting Pt: 1340 °C Density: 2.564 Storage Temperature: Ambient |
MDL Number:
MFCD00036295 CAS Number: 7778-53-2 EINECS: 231-907-1 Merck Index: 14,7660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...