Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
35926-KT 35947-KT 35968-KT 36039-KT 36058-KT 36083-KT
CAAA35926-KTEA 178.54 CAD
CAAA35926-KT CAAA35947-KT CAAA35968-KT CAAA36039-KT CAAA36058-KT CAAA36083-KT
Low Form Zirconium Crucibles, Thermo Scientific
Crucibles
Zirconium crucibles are handmade to an exacting tolerance for uniform wall thickness
  • Special apparatus is not required for handling hot zirconium crucibles
  • Sudden contact with cold, metallic surfaces will have no deleterious effect on a zirconium crucible
  • Zirconium crucibles require a minimum of specialized care; smoothing and shaping is not a special consideration
  • The inherent strength of zirconium precludes the necessity of reinforced rims and thicker bottoms
These crucibles are produced under the most stringent requirements to ensure ultimate tensile strength, yield strength, elongation, and chemical purity.
Only high-purity zirconium material is used.