Samarium diiodide 0.07-0.12 M THF Stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Samarium (II) iodide

022360-AE 022360-AP
AA22360-AEEA 159.58 CAD
AA22360-AE AA22360-AP
Samarium diiodide 0.07-0.12 M THF Stabilised
Samarium diiodide

CAS No.: 32248-43-4 Molecular Formula: SmI2 Formula Weight: 404.21 Physical Form: Liquid Density: 0.922 Sensitivity: Air and Moisture Sensitive UN No.: 3271 MDL No.: MFCD00058873

Formula: I₂Sm
MW: 404.17
Density: 0.922
Storage Temperature: Ambient
MDL Number: MFCD00058873
CAS Number: 32248-43-4
EINECS: 203-726-8
UN: 2056
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR