Potassium chloride, dry_ultra 99,95
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Potassiumchloride
14466-09
14466-18
14466-30
CAAA14466-09EA
135.09
CAD
CAAA14466-09
CAAA14466-18
CAAA14466-30
Potassium chloride, dry_ultra 99,95
Potassium chloride
Purity: 99.95%
CAS No.: 7447-40-7
Molecular Formula: KCl
Formula Weight: 74.55
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 1.984
Refractive Index: 1.490
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 773°
Boiling Point: 1500° subl.
Merck Reference: 14,7621
MDL No.: MFCD00011360
Formula:
KCl MW: 74.55 g/mol Boiling Pt: 1420 (subl.) Melting Pt: 773 Density: 1.984 Storage Temperature: Ambient |
MDL Number:
MFCD00011360 CAS Number: 7447-40-7 EINECS: 231-211-8 Merck Index: 14,07621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...