Cobalt ≥99.995% (metals basis), rods, Puratronic®, Ø 12 mm (0.47 in)

Supplier: Thermo Scientific Chemicals
Danger

10867-DJ 10867-HF
CAAA10867-DJEA 2101.4 CAD
CAAA10867-DJ CAAA10867-HF
Cobalt ≥99.995% (metals basis), rods, Puratronic®, Ø 12 mm (0.47 in)
Cobalt

Purity: 99.995% CAS No.: 7440-48-4 Physical Form: ≈10.1g/cm Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 2809
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR