Adson Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
50-1372 50-1472
89025-056EA 310.53 CAD
89025-056 89025-058
Adson Hemostatic Forceps, OR Grade, Sklar
Forceps
Adson Hemostatic Forceps are used for clamping small vessels to halt blood flow, particularly with the tonsils.
The jaws of these Adson forceps are 1/4 the length of the shanks and feature horizontal serrations half the length of the jaw. The shanks are longer than both Crile and Kelly forceps.