VWR® Water Jacketed CO₂ Incubators
Supplier: VWR International
10810-744 10810-878 10810-884 10810-886
10810-744EA 13077.54 CAD
10810-744 10810-878 10810-884 10810-886
VWR® Water Jacketed CO₂ Incubators
Incubators CO₂ Incubators
The VWR® Water Jacketed CO2 incubator protects your cultures from unwanted heat loss by surrounding them with triple-walled construction and a jacket of temperature-conditioned water.
  • 6.5 cu. ft. internal chamber volume
  • Supplied with four removable stainless steel shelves and water pan
  • Outstanding temperature stability and security
  • Fan-assisted airflow and in-chamber sensors maintain a uniform and stable growth environment
  • Choice of long-lasting TC or independent IR CO2 sensors
  • Dual temperatures for over-temperature protection
  • Easy assembly and disassembly
More Product Information