PARition Wall Systems Packages, Quantum Storage Systems
Supplier: QUANTUM STORAGE SYSTEMS
WS70-SS18-7S WS70-SS36-7S WS70-SS36-4S3L
10805-756EA 3130.34 CAD
10805-756 10805-760 10805-952
PARition Wall Systems Packages, Quantum Storage Systems
Furniture Shelves
PARtition wall hanging baskets are uniquely designed for instant visibility and easy access while maximizing storage area.
  • The wire construction minimizes dirt accumulation and allows air circulation
  • Chrome finish
Available in ⁹⁵/₈" and ¹⁴³/₈" width and in 18" and 36" lengths.
Baskets have a ¹/₂×¹/₂" grid pattern to partition products while letting you know when to reorder product and restock items.
Baskets 11 7/8 x 18 x 7 1/2"; 11 7/8 x 36 x 7 1/2"