Gerald Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
47-1173 47-1186
89024-942EA 203.41 CAD
89024-942 89024-948
Gerald Tissue Forceps, OR Grade, Sklar
Forceps
Sklar® Gerald Tissue Forceps are a light to intermediate weight instrument with very narrow tips specifically used to handle delicate tissue
  • They feature 1 x 2 teeth to securely grasp the tissue, but also have a stop peg to prevent an overly harsh grasp that may crush the tissue.
  • Censitrac Ready OR Grade Gerald Tissue Forceps are industry standard and constructed of stainless steel.
They are often used in cardiothoracic procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.