Econo™ Sterile Tissue Forceps, Sklar
Supplier: Sklar
96-2578 96-2579 96-2661 96-2574
89023-900CS 370.46 CAD
89023-900 89023-902 89023-980 75995-408
Econo™ Sterile Tissue Forceps, Sklar
Forceps
Econo™ Sterile Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures.
  • Spring tension shanks determine how far instrument will open
  • Toothed to grip tissue more firmly than serrated tips alone
  • Eliminates the need for cleaning and reprocessing
  • Significantly reduce likelihood of cross-contamination
These instruments have spring tension shanks that determine how far the instrument will open and the thickness of the tissue that can be held
Econo™ Sterile instruments are ready-to-use, sterile,More Product Information