Econo™ Sterile Halsted Mosquito Forceps, Floor Grade, Sklar
Supplier: Sklar
96-2657 96-2658
89023-972CS 941.15 CAD
89023-972 89023-974
Econo™ Sterile Halsted Mosquito Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Halsted Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels.
The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate.
Order Now
SPECIFICATIONS
Environmentally Preferable