Hoke Osteotome, OR Grade, Sklar
Supplier: Sklar
40-6953 40-6954 40-6956 40-6958 40-6970 40-6973 40-6976 40-6979
10196-172EA 253.1 CAD
10196-172 10196-174 10196-176 10196-178 10196-180 10196-182 10196-184 10196-186
Hoke Osteotome, OR Grade, Sklar
Chisels
Sklar's® Hoke Osteotome is used for cutting and reshaping bony areas.
  • Straight or curved with different tip sizes
Typically smaller than the Hibbs osteotome, the Hoke osteotome is often used in smaller surgical sites.