Econo™ Sterile Adson Tissue Forceps, Sklar
Supplier: Sklar
96-2572 96-2573 96-2656 96-2775
89023-890CS 275.78 CAD
89023-890 89023-892 89023-970 75995-428
Econo™ Sterile Adson Tissue Forceps, Sklar
Forceps
Econo™ Sterile Adson Tissue Forceps are for holding and manipulating delicate tissues.
  • Wide, flat thumb grasp area
  • Short jaws and narrow tips
  • Ready-to-use
The forceps are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities.