Mixter Scissors, OR Grade, Sklar
Supplier: Sklar
15-1360 15-1361
89025-496EA 246.11 CAD
89025-496 76191-666
Mixter Scissors, OR Grade, Sklar
Scissors
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar can deliver superior precision for any surgical procedure
Mixter Scissors are medium weight scissors used for dissection of medium weight tissue. Available both straight and curved, the jaws of Mixter scissors are 1/2 the length of the shanks.