Sklar Edge™ Tungsten Carbide Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
16-1805 16-1905 16-2005 16-2105 16-2115 16-3503 16-3513 16-3514 16-3506 16-1815 16-3526 16-3527
89025-680EA 654.14 CAD
89025-680 89025-684 89025-688 89025-690 89025-694 89025-698 76191-988 76192-014 76192-016 76192-018 76192-048 76192-060 76192-062
Sklar Edge™ Tungsten Carbide Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors.
Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.