Sklar Edge™ Tungsten Carbide Operating Scissors, OR Grade, Sklar
Supplier: Sklar
16-1055 16-1056 16-1057 16-1155 16-1156 16-1157
89025-656EA 596.19 CAD
89025-656 89025-658 89025-660 89025-662 89025-664 89025-666
Sklar Edge™ Tungsten Carbide Operating Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument
TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting.