Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
22-2450 22-2460
89025-282EA 270.46 CAD
89025-282 89025-284
Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Scissors
Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.
They are not recommended for cutting sutures, drains, or heavy tissue.