Solvent Reservoirs, Plastic Coated, Chemglass
Supplier: Chemglass
CG-1166-02 CG-1166-03 CG-1166-04 CG-1166-05
80060-484EA 1152.16 CAD
80060-484 80060-486 80060-488 80060-490
Solvent Reservoirs, Plastic Coated, Chemglass
Bottles Solution Bottles
Complete reservoir system includes a plastic-coated graduated bottle, a standard cap assembly, solvent and sparging filters.
  • High performance
  • Low pressure liquid chromatography mobile phase
Cap assembly fits GL45 threaded bottles and has a PTFE bulkhead with three ¹/₄"-28 threaded ports to accommodate fittings for ¹/₈" Ext.Ø tubing.