Ribbon Utility Scissors, OR Grade, Sklar
Supplier: Sklar
47-1146 47-1247
89024-940EA 197.26 CAD
89024-940 89024-950
Ribbon Utility Scissors, OR Grade, Sklar
Scissors
Ribbon Utility Scissors are a multipurpose instrument used for cutting lightweight, non-sterile materials such as gauze, bandages and dressing material.
This product is straight with smooth edges, and ribbon style handles for additional comfort.
Angular Scissors are finger ring scissors comparable to iris scissors.