Anti-SOX2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 20G5]

Supplier: Pierce Biotechnology
MA1020D488
CAPIMA1020D488EA 2065.52 CAD
CAPIMA1020D488
Anti-SOX2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 20G5]
Antibodies
This Antibody has been successfully used in Western blot with human and mouse samples. Western blot analysis of MA1-014-HRP specifically detects SOX2 protein at ~36 kDa in the nucleus of embryonal carcinoma cells. This intronless gene encodes a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of this gene is required for stem-cell maintenance in the central nervous system, and also regulates gene expression in the stomach. Mutations in this gene have been associated with optic nerve hypoplasia and with syndromic microphthalmia, a severe form of structural eye malformation. This gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT).

Recommended Dilutions: Western Blot: 1:250-1:1000

Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: 20G5
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR