Surgi-OR™ Kelly Hemostatic Forceps, Mid Grade, Sklar
Supplier: Sklar
95-436 95-442
89023-618EA 62.97 CAD
89023-618 89023-620
Surgi-OR™ Kelly Hemostatic Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Kelly Hemostatic Forceps are used for clamping large blood vessels or manipulating heavy tissue
They are available with a ratcheted finger ring handle. Similar to Rankin forceps, the jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Kelly forceps are shorter and heavier than Rankin forceps.
Surgi-OR™ products are mid-grade instruments suitable for the operating room and other surgical centers.
They may be used for soft tissue dissection.