Sklar Black™ Kogan Endocervical Speculum, OR Grade, Sklar
Supplier: Sklar
90-8850 90-8855 90-8858 90-8860 90-8862 90-8865
10199-174EA 1931.94 CAD
10199-174 10199-176 10199-178 10199-180 10199-182 10199-184
Sklar Black™ Kogan Endocervical Speculum, OR Grade, Sklar
Specula
Sklar Black™ Kogan laser endocervical speculum is a gynecological instrument used for viewing the distal endocervical canal.
Unlike a traditional, duckbill speculum, this single spring, locking, finger-ring intrument features opposing, fenestrated blades at the end of long jaws. The jaws and blades extend deeper into the cervix and can therefore retract deeper tissue. An endocervical speculum is also used in colposcopy procedures, or where polyps cannot be easily visualized.
Sklar Black™ laser instruments feature a non-reflective black polymer coating applied over the sandblastedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable