Sklar Black™ Frazier Tissue Hook, OR Grade, Sklar
Supplier: Sklar
90-1048 90-1049
10199-052EA 454.03 CAD
10199-052 10199-054
Sklar Black™ Frazier Tissue Hook, OR Grade, Sklar
Retractors
Sklar Black™ Frazier tissue hook is a single-prong instrument used for gentle handling and retracting of tissue, commonly in dermatologic procedures.
  • Straight or curved
These instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.
This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: Comply with Biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.