Sklar Black™ Emmett Hook, Quick Release, OR Grade, Sklar
Supplier: Sklar
90-2114 90-2116 90-2118 90-2120 90-2122 90-2124 90-2126
10197-180EA 474.43 CAD
10197-180 10197-182 10197-184 10197-186 10197-188 10197-190 10197-192
Sklar Black™ Emmett Hook, Quick Release, OR Grade, Sklar
Retractors
The Sklar Black™ emmett hook, which may also be known as an emmett iris hook, is most commonly used in OB/GYN procedures involving the uterus.
  • Safe and durable coating
  • Reduces laser reflection to less than one-tenth of a percent
The hook, available in multiple diameters, is at the end of a long, narrow handle, often 9 or 12 inches in length. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries.