Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
T3002 T3001
76020-660EA 307.2 CAD
76020-660 77002-226
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Nucleic Acid Reagents Transformation Reagents
The Mix & Go!™ E. coli transformation kit and Mix & Go!™ E. coli transformation buffer set are convenient methods for the preparation of competent E. coli cells for simple and highly efficient DNA transformation.
  • It is very simple and efficient DNA transformation
  • It completely eliminates the requirement for heat shocking and related procedures
  • Transformation efficiencies are typically on the order of 10to 109 transformants/µg plasmid DNA
The Mix & Go! method completely eliminates the requirement for heat shocking and related procedures. Instead, Mix & Go! bacterial transformation can be performed by adding DNA to Mix & Go! competent cells and spreading onto a plate. Transformation efficiencies are typically on the order of 10to 109 transformants/µg plasmid DNA with most E. coli strains. These formulated reagents make it easy to generate Mix & Go! competent cells from current E. coli strains that are available in the laboratory. Simply grow the E. coli strain of your choice, wash, then resuspend the cells in the provided buffers. The cells are now transformation ready. The Mix & Go! E. coli Transformation Kit includes all buffers and ZymoBroth medium to generate 20 ml of Mix & Go! competent cells. The Mix & Go! E. coli Transformation Buffer Set includes all buffers that are required to generate 60 ml of Mix & Go! competent cells, and the medium (broth) is supplied by the user.