Zymoclean™ Gel DNA Recovery Kits, Zymo Research

Supplier: Zymo Research

D4001 D4002 D4007 D4008 D4021 D4022
77001-116EA 226.71 CAD
77001-116 77001-122 77001-144 77001-146 77001-168 77001-170
Zymoclean™ Gel DNA Recovery Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents

The Zymoclean Gel DNA Recovery Kits provide for the rapid purification of high quality DNA from TAE/TBE-bufffered agarose gels.


  • Quick (15 minute) recovery of ultra-pure DNA from agarose gels.
  • Column design permits DNA elution at high concentrations into minimal volumes (≥ 6 µL).
  • Eluted DNA is well suited for use in DNA ligation, sequencing, labeling, PCR, etc.


About Zymoclean™ Gel DNA Recovery Kit


The Zymoclean™ Gel DNA Recovery Kit provides for the rapid purification of high quality DNA from TAE/TBE-buffered agarose gels. The product features Fast-Spin technology to yield high-quality, purified DNA in just minutes. DNA purified using the Zymoclean™ Gel DNA Recovery kit are perfectly suited for use in DNA ligation reactions, sequencing, DNA labeling reactions, PCR, etc.

Order Now

SPECIFICATIONS

Description Zymoclean™ Gel DNA Recovery Kit ZR-96 Zymoclean™ Gel DNA Recovery Kit
Environmentally Preferable
Application
Target molecule DNA DNA
Refine target molecule DNA gel bands DNA gel bands
Sample type Gel matrix Gel matrix
Method / Format Column (silica-based) Multiwell plate
Prep size Mini Mini
Yield 50 bp to 23 kb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR