YMC-Pack ODS-AQ HPLC Columns, YMC America
Supplier: YMC America,Inc
AQ20S03-1002WT AQ20S03-1046WT AQ12S05-1010WT AQ12S03-0502WT AQ12S03-0503WT AQ12S03-0504WT AQ12S03-0546WT AQ12S03-1002WT AQ12S03-1003WT AQ12S03-1006WT AQ12S03-1046WT AQ12S03-1502WT AQ12S03-1503WT AQ12S03-1506WT AQ12S03-1546WT AQ12S03-2502WT AQ12S03-2503WT AQ12S03-2546WT AQ12S03-H502WT AQ12S03-L502WT AQ12S05-0502WT AQ12S05-0546WT AQ12S05-1002WT AQ12S05-1006WT AQ12S05-1020WT AQ12S05-1030WT AQ12S05-1046WT AQ12S05-1501WT AQ12S05-1502WT AQ12S05-1503WT AQ12S05-1506WT AQ12S05-1510WT AQ12S05-1520WT AQ12S05-1530WT AQ12S05-1546WT AQ12S05-2501WT AQ12S05-2502WT AQ12S05-2503WT AQ12S05-2506WT AQ12S05-2510WT AQ12S05-2520WT AQ12S05-2530WT AQ12S05-2546WT AQ12S05-3006WT AQ12S05-3010WT AQ12S05-3046WT AQ12S05-L502WT AQ12S05-L530WT AQ12S05-L546WT AQ20S03-0501WT AQ20S03-1502WT AQ20S03-1546WT AQ20S05-1002WT AQ20S05-1520WT AQ20S05-1546WT AQ20S05-2502WT AQ20S05-2503WT AQ20S05-2520WT AQ20S05-2530WT AQ20S05-2546WT AQ12S05-0503WT AQ12S05-0504WT AQ20S03-0502WT AQ20S03-0503WT AQ20S03-0504WT AQ20S05-0502WT AQ20S05-0503WT AQ20S05-0504WT AQ20S05-1046WT
89198-924EA 1964.86 CAD
89198-924 89198-926 89198-932 97028-856 97028-860 97028-864 97028-868 97028-870 97028-872 97028-874 97028-878 97028-880 97028-882 97028-884 97028-886 97028-888 97028-890 97028-892 97028-902 97028-904 97028-928 97028-942 97028-946 97028-948 97028-950 97028-954 97028-958 97028-960 97028-962 97028-964 97028-966 97028-968 97028-970 97028-972 97028-974 97028-976 97028-978 97028-980 97028-982 97028-984 97028-986 97028-988 97028-990 97028-992 97028-996 97028-998 97029-000 97029-002 97029-012 97029-014 97029-016 97029-118 97029-128 97029-130 97029-154 97029-158 97029-160 97029-162 97029-164 97029-166 97029-168 97029-170 89216-474 89216-476 89216-486 89216-488 89216-490 89216-498 89216-500 89216-502 89216-504 10811-194 10811-196 10811-234
YMC-Pack ODS-AQ HPLC Columns, YMC America
Chromatography Columns
ODS-AQ is a unique reversed-phase material with both a hydrophobic high carbon loading and a relatively hydrophilic surface.
  • Unique, reverse-phased material
  • Rproducible retention
  • Hydrophillic surface
ODS-AQ provides chromatographers reproducible retention and excellent peak shape in highly aqueous eluents; even 100% water.