Sklar Black™ Graves Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-9015 90-9054 90-9016 90-9018 90-9020 90-3804 90-3806 90-3808 90-3810 90-3812
10184-260EA 830.15 CAD
10184-260 10184-272 10194-008 10194-096 10194-098 10197-216 10197-218 10197-220 10197-222 10197-224
Sklar Black™ Graves Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Black™ Graves Vaginal Speculum is used for laser gynecological procedures.
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.
Delivery information: The Sklar Black™ Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation.